Learn More
Alkali Scientific Alkali Scientific™ C5000-8-PT Sprint™ 8 Portable Clinical Centrifuge
SDP
Catalog No. 503029543
Encompass_Preferred
Alkali C5000-8-PT Sprint™8 Portable Clinical Centrifuge The Sprint 8 includes an eight position, fixed-angle rotor for the centrifugation of 10ml blood tubes (16 x 100mm), and 15ml conical tubes (17 x 120mm). Smaller blood tubes (12 x 75mm and 12 x 100mm) are also compatible with the use of optional adapters (C3100-ADP). With intuitive controls and a large LED display, virtually no programming or setup is required prior to starting a cycle. Simply use the up/down buttons to select the run time and press “START”. The safety lid-lock is engaged and the rotor quickly and smoothly accelerates to 3,400 rpm. Once the run time is completed (or the “STOP” button is pressed) the rotor comes to a complete stop and the lid lock is disengaged for the removal of samples
Catalog No. 50-302-9543 Supplier Alkali Scientific Supplier No. RS75361
Alkali C5000-8-PT Sprint™8 Portable Clinical Centrifuge The Sprint 8 includes an eight position, fixed-angle rotor for the centrifugation of 10ml blood tubes (16 x 100mm), and 15ml conical tubes (17 x 120mm). Smaller blood tubes (12 x 75mm and 12 x 100mm) are also compatible with the use of optional adapters (C3100-ADP). With intuitive controls and a large LED display, virtually no programming or setup is required prior to starting a cycle. Simply use the up/down buttons to select the run time and press “START”. The safety lid-lock is engaged and the rotor quickly and smoothly accelerates to 3,400 rpm. Once the run time is completed (or the “STOP” button is pressed) the rotor comes to a complete stop and the lid lock is disengaged for the removal of samples