Learn More
Alkali Scientific Alkali Scientific™ C5000-8 Sprint™ 8 Clinical Centrifuge with 8 x 15ml fixed angle rotor, 115V
SDP
Catalog No. 503029514
Encompass_Preferred
8 Position fixed angle rotor Holds common research and blood collection tubes up to 15ml Fixed at 3,400rpm, ideal for most coag, plasma (PPP) and urinalysis applications Adjustable timer The Alkali Scientific™ C5000-8 Sprint™ 8 includes an eight position, fixed angle rotor for the centrifugation of 10ml blood tubes (16 x 100mm), and 15ml conical tubes (17 x 120mm). Smaller blood tubes (12 x 75mm and 12 x 100mm) are also compatible with the use of optional adapters (C3100-ADP).With intuitive controls and a large LED display, virtually no programming or setup is required prior to starting a cycle. Simply use the up/down buttons to select the run time and press “START”. The safety lid-lock is engaged and the rotor quickly and smoothly accelerates to 3,400 rpm. Once the run time is completed (or the “STOP” button is pressed) the rotor comes to a complete stop and the lid lock is disengaged for removal of samples. Ideal for Coag, PPP and urinalysis studies.
Catalog No. 50-302-9514 Supplier Alkali Scientific Supplier No. RS7534
8 Position fixed angle rotor Holds common research and blood collection tubes up to 15ml Fixed at 3,400rpm, ideal for most coag, plasma (PPP) and urinalysis applications Adjustable timer The Alkali Scientific™ C5000-8 Sprint™ 8 includes an eight position, fixed angle rotor for the centrifugation of 10ml blood tubes (16 x 100mm), and 15ml conical tubes (17 x 120mm). Smaller blood tubes (12 x 75mm and 12 x 100mm) are also compatible with the use of optional adapters (C3100-ADP).With intuitive controls and a large LED display, virtually no programming or setup is required prior to starting a cycle. Simply use the up/down buttons to select the run time and press “START”. The safety lid-lock is engaged and the rotor quickly and smoothly accelerates to 3,400 rpm. Once the run time is completed (or the “STOP” button is pressed) the rotor comes to a complete stop and the lid lock is disengaged for removal of samples. Ideal for Coag, PPP and urinalysis studies.