Learn More
Strem, An Ascensus Company CAS# 51364-51-3. Tris(dibenzylideneacetone)dipalladium(0). MFCD00013310. 25G
SDP
Supplier:  Strem, An Ascensus Company 46300025G
Encompass
CAS# 51364-51-3. 25G. Tris(dibenzylideneacetone)dipalladium(0). MFCD00013310. Molecular Formula: C51H42O3Pd2. Molecular Weight: 915.70. Color/Form: Stability: air sensitive, moisture sensitive. Strem# 46-3000. www.strem.com/catalog/v3/46-3000/
Catalog No. 50-195-9497