Learn More
iQ Biosciences C57BL/6 Mouse Irradiated Splenocytes, 25M cells/vial
Catalog No. NC1996993
Encompass_Preferred
High-quality, high-viability (typically > 70%) irradiated splenocytes from the C57BL/6 mouse, the most commonly-used inbred mouse strain. Used for a wide variety of immunology-based applications, including proliferation and cytotoxicity studies. All orders come with an iQ Certificate of Analysis.
Catalog No. NC1996993 Supplier iQ Biosciences Supplier No. IQBM2S25R
High-quality, high-viability (typically > 70%) irradiated splenocytes from the C57BL/6 mouse, the most commonly-used inbred mouse strain. Used for a wide variety of immunology-based applications, including proliferation and cytotoxicity studies. All orders come with an iQ Certificate of Analysis.