Learn More
Cell Biologics C57BL/6 MOUSE LUNG MACROPHAGES
SDP
Catalog No. NC1947458
Encompass_Preferred
C57BL/6 MOUSE LUNG MACROPHAGES (FROZEN CELLS) C57-2313FC57BL/6 Mouse Lung Macrophages are isolated from the lung of pathogen-free laboratory adult mice. Cells at passage 0 are cryo-preserved in vials. Each vial contains 0.5x106 cells and is delivered frozen. Cells are negative for bacteria, yeast, fungi, and mycoplasma and tested for expression of markers using antibody, CD11b by flow cytometry. Cells can be expanded on a multiwell culture plate ready for experiments under the cell culture conditions specified by Cell Biologics. Repeated freezing and thawing of cells is not recommended.
Catalog No. NC1947458 Supplier Cell Biologics Supplier No. C572313F
C57BL/6 MOUSE LUNG MACROPHAGES (FROZEN CELLS) C57-2313FC57BL/6 Mouse Lung Macrophages are isolated from the lung of pathogen-free laboratory adult mice. Cells at passage 0 are cryo-preserved in vials. Each vial contains 0.5x106 cells and is delivered frozen. Cells are negative for bacteria, yeast, fungi, and mycoplasma and tested for expression of markers using antibody, CD11b by flow cytometry. Cells can be expanded on a multiwell culture plate ready for experiments under the cell culture conditions specified by Cell Biologics. Repeated freezing and thawing of cells is not recommended.