Learn More
iQ Biosciences C57BL/6 Mouse PBMCs, 5M cells/vial
Catalog No. NC0853050
Encompass_Preferred
High-quality, high-viability (> 80%) peripheral blood mononuclear cells (PBMCs) from the C57BL/6 mouse, the most commonly-used inbred mouse strain. Used for a wide variety of immunology-based applications, including in-vitro immunoassays and ex-vivo applications. All orders come with an iQ Certificate of Analysis.
Catalog No. NC0853050 Supplier iQ Biosciences Supplier No. IQBMPB201
High-quality, high-viability (> 80%) peripheral blood mononuclear cells (PBMCs) from the C57BL/6 mouse, the most commonly-used inbred mouse strain. Used for a wide variety of immunology-based applications, including in-vitro immunoassays and ex-vivo applications. All orders come with an iQ Certificate of Analysis.