Learn More
Cyagen Biosciences Inc C57BL/6 Mouse Bone Marrow Mesenchymal Stem Cells
SDP
Catalog No. NC2960401
Encompass
OriCellTM C57BL/6 mouse bone marrow mesenchymal stem cells are extracted from healthy C57BL/6 mouse bone marrow and have strong proliferation and multi-directional differentiation capabilities. It can be used as a cell model to study proliferation, aging, immunity, differentiation and transplantation.
Catalog No. NC2960401 Supplier Cyagen Biosciences Inc Supplier No. MUBMX01001
OriCellTM C57BL/6 mouse bone marrow mesenchymal stem cells are extracted from healthy C57BL/6 mouse bone marrow and have strong proliferation and multi-directional differentiation capabilities. It can be used as a cell model to study proliferation, aging, immunity, differentiation and transplantation.