Learn More
INTACT GENOMICS INC C58C1 Agrobacterium Chemically Competent Cells 18x50µl
Catalog No. 5000083057
Encompass
Intact Genomics (ig) C58C1 Agrobacterium Chemically Competent Cells are optimized for the highest transformation efficiency and are useful for various applications. The chromosomal background of C58C1 is C58. C58 is cured of the Ti plasmid pTiC58 resulting in C58C1. C58C1 Competent cells may be useful for transgenic operations that involve Arabidopsis and other plants. This Agrobacterium strain is streptomycin and rifampicin-resistant.Useful for transgenic operations that involve potatoes, astragalus, rosaceae, tobacco and other plants.Product Includes and Storage: C58C1 Agrobacterium Chemically Competent Cells: -80 ºCpCAMBIA1391z control DNA: -20 ºCAgro Recovery medium: 4 ºC
Catalog No. 50-000-83057 Supplier INTACT GENOMICS INC Supplier No. 108618
Intact Genomics (ig) C58C1 Agrobacterium Chemically Competent Cells are optimized for the highest transformation efficiency and are useful for various applications. The chromosomal background of C58C1 is C58. C58 is cured of the Ti plasmid pTiC58 resulting in C58C1. C58C1 Competent cells may be useful for transgenic operations that involve Arabidopsis and other plants. This Agrobacterium strain is streptomycin and rifampicin-resistant.Useful for transgenic operations that involve potatoes, astragalus, rosaceae, tobacco and other plants.Product Includes and Storage: C58C1 Agrobacterium Chemically Competent Cells: -80 ºCpCAMBIA1391z control DNA: -20 ºCAgro Recovery medium: 4 ºC