Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Complement Technology Inc C5a Anaphylatoxin, 30 μg/vial, Frozen liquid

Supplier:  Complement Technology Inc A144

Encompass

Human C5a (Anaphylatoxin) is a highly potent polypeptide essential for regulating the complement system. This product, derived from human serum, is enzymatically processed from human C5 protein and contains 74 amino acids. It serves various biological roles, including chemotactic activity for neutrophils and facilitating smooth muscle contraction, vascular permeability, and histamine release. Supplied as a 30 μg vial of frozen liquid, it is renowned for its purity and activity, catering to advanced research needs in immunology and complement studies.

  • Source is normal human serum
  • Form is a frozen liquid
  • Vial size is 30 μg
  • Polypeptide length is 74 amino acids
  • Purity exceeds 97% by SDS-PAGE
  • Typically < 0.1 EU/μg endotoxin level
  • Stored at -70°C or below
  • Manufactured in the USA
  • Research use only

Catalog No. NC1730157


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.