Learn More
Aviva Systems Biology Corp C5AR1 Antibody - C-terminal region (ARP60075_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP60075_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against C5AR1. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human C5AR1. Alternate names: C5A, C5AR, C5R1, CD88ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Sheep
Catalog No. 50-286-934