Learn More
Eurofins Discoverx PathHunter C5AR1 Bioassay Kit, 10-Plate. (93-0557Y2-00060)
Catalog No. 502000791
Encompass
The PathHunter C5AR1 Bioassay kit provides an easy-to-use cell based assay to measure drug potency and detect neutralizing antibodies. This bioassay assesses ligand (e.g. C5a) based activation of C5AR1 GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged C5AR1 and EA-tagged β-Arrestin-2. Activation of C5AR1 stimulates the recruitment of β-Arrestin-2 and produces EFC signal. Bioassay kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, control agonist, detection reagent, and assay plates. For detailed product information, User manual and COA, visit www.discoverx.com/bioassay.
Catalog No. 50-200-0791 Supplier Eurofins Discoverx Supplier No. 930557Y200060
The PathHunter C5AR1 Bioassay kit provides an easy-to-use cell based assay to measure drug potency and detect neutralizing antibodies. This bioassay assesses ligand (e.g. C5a) based activation of C5AR1 GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged C5AR1 and EA-tagged β-Arrestin-2. Activation of C5AR1 stimulates the recruitment of β-Arrestin-2 and produces EFC signal. Bioassay kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, control agonist, detection reagent, and assay plates. For detailed product information, User manual and COA, visit www.discoverx.com/bioassay.