Learn More
Enzo Life Sciences Complement component C5b-9 monoclonal antibody (aE11) (1 mg)
SDP
Supplier:  Enzo Life Sciences BPDDIA011011
Encompass_Preferred
Clone: aE11. Isotype: IgG2aκ. Immunogen: Purified human complement component C5b-9. Species reactivity: Human Porcine, Horse, Baboon. Applications: ELISA, IF, IHC. Purity: Protein A-affinity purified. Formulation: Liquid. In 10mM phosphate buffer, pH 7.4, containing 0.15M sodium chloride and 0.09% sodium azide.
Catalog No. 50-201-2452