Learn More
Aviva Systems Biology Corp C5orf35 Antibody - N-terminal region (ARP53194_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP53194_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against C5orf35. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human C5orf35. Alternate names: MGC33648, C5orf35ApplicationWB. Species reactivity: Human
Catalog No. 50-014-60965