Learn More
Aviva Systems Biology Corp C5orf36 Antibody - N-terminal region (ARP55682_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP55682_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against C5orf36. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human C5orf36. Alternate names: DKFZp686F0372, MGC34713, C5orf36ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Pig, Rabbit, Rat, Yeast
Catalog No. 50-014-61576