Learn More
Aviva Systems Biology Corp C5orf4 Antibody - N-terminal region (ARP42675_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42675_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against C5orf4. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human C5orf4. Alternate names: FLJ13758, C5orf4ApplicationIHC, WB. Species reactivity: Dog, Guinea Pig, Horse, Human, Pig, Rabbit, Rat
Catalog No. 50-014-58301