Learn More
Cayman Chemical C6 Biotin Ceramide (d18:1/6:0), Purity: ≥98%, CAS: 192070-02-3, 5 mg
SDP
Catalog No. 502895320
Encompass_Preferred
An affinity probe for C6 ceramide binding partners
Catalog No. 50-289-5320 Supplier Cayman Chemical Supplier No. 90004155 MG
An affinity probe for C6 ceramide binding partners