Learn More
Cayman Chemical C6 Biotin Ganglioside GM1 d1
SDP
Catalog No. 503016040
Encompass_Preferred
An affinity probe for ganglioside GM1 binding partners; has been used to detect ganglioside GM1 binding partners in vitro
Catalog No. 50-301-6040 Supplier Cayman Chemical Supplier No. 24840500 UG
An affinity probe for ganglioside GM1 binding partners; has been used to detect ganglioside GM1 binding partners in vitro