Learn More
Cayman Chemical C6 CeramIded18 16 0 Alkyn 5mg
SDP
Catalog No. 503015688
Encompass_Preferred
A geranylgeranyl transferase I inhibitor (IC50 38 nM) 140-fold selective for GGTase I over farnesyltransferase (IC50 5400 nM) inhibits geranylgeranylation of RAP1A in vitro (IC50 10 UM) inhibits cell growth and decreases migration and invasion of OSSC cells to 75 45 and 27 of controls respectively prevents ovalbumin-induced eosinophil infiltration into airways in a mouse model of allergic bronchial asthma (5 mg/kg per day i.p.) decreases morphine withdrawal severity in rats (ED50 0.076 mg/kg)
Catalog No. 50-301-5688 Supplier Cayman Chemical Supplier No. 245195 MG
A geranylgeranyl transferase I inhibitor (IC50 38 nM) 140-fold selective for GGTase I over farnesyltransferase (IC50 5400 nM) inhibits geranylgeranylation of RAP1A in vitro (IC50 10 UM) inhibits cell growth and decreases migration and invasion of OSSC cells to 75 45 and 27 of controls respectively prevents ovalbumin-induced eosinophil infiltration into airways in a mouse model of allergic bronchial asthma (5 mg/kg per day i.p.) decreases morphine withdrawal severity in rats (ED50 0.076 mg/kg)