Learn More
Cayman Chemical C6 CeramIded7d18 1d76 0 100ug
SDP
Catalog No. 503015991
Encompass_Preferred
An internal standard for the quantification of short-chain ceramides by GC- or LC-MS
Catalog No. 50-301-5991 Supplier Cayman Chemical Supplier No. 24826100 UG
An internal standard for the quantification of short-chain ceramides by GC- or LC-MS