Learn More
Enzo Life Sciences C6 Dihydroceramide (5 mg). CAS: 171039-13-7
SDP
Catalog No. 502012106
Encompass_Preferred
C6 Dihydroceramide is inactive and may be used as a negative control for C6 ceramide. Purity: ≥98%. Solubility: Soluble in DMF (25mg/ml), DMSO (5mg/ml) or ethanol (10mg/ml). Long Term Storage: -20°C.
Catalog No. 50-201-2106 Supplier Enzo Life Sciences Supplier No. BMLSL1110005
C6 Dihydroceramide is inactive and may be used as a negative control for C6 ceramide. Purity: ≥98%. Solubility: Soluble in DMF (25mg/ml), DMSO (5mg/ml) or ethanol (10mg/ml). Long Term Storage: -20°C.