Learn More
AdipoGen C6-DL-HSL
SDP
Supplier:  AdipoGen CDXH0200M025
Encompass_Preferred
Chemical. CAS 106983-28-2. Formula C10H17NO3. MW 199.3. Synthetic. N-Hexanoyl-DL-homoserine lactone is a small diffusible signaling molecule involved in quorum sensing, controlling gene expression and affecting cellular metabolism. The diverse applications of this kind of molecule include regulation of virulence in general and in cystic fibrosis, infection prevention, slime and biofilm reduction in commercial agriculture and aquaculture industries, food spoilage prevention and septicemia in fish.
Catalog No. 50-256-0706