Learn More
ATCC C6; Glioma; Rat (Rattus novegicus)
Catalog No. 502383114
Encompass
Due to an upcoming ERP system upgrade, ATCC will have a temporary system shutdown from 11/20/26-12/1/26. All orders will experience a 5-6 days delay in processing. Orders will continue to be sent to ATCC during the blackout and will be held until 12/2/26 when they are released. Any questions, please reach out to Encompass.Supplierchannel@thermofisher.com.
C6 is a glial cell line that was isolated from the brain of a rat with Glioma. The glial tumor was induced by N-nitrosomethylurea after a series of alternate culture and animal passages. This cell line was deposited by G. Sato and can be used in neuroscience and toxicology research.
Catalog No. 50-238-3114 Supplier ATCC Supplier No. CCL107
Due to an upcoming ERP system upgrade, ATCC will have a temporary system shutdown from 11/20/26-12/1/26. All orders will experience a 5-6 days delay in processing. Orders will continue to be sent to ATCC during the blackout and will be held until 12/2/26 when they are released. Any questions, please reach out to Encompass.Supplierchannel@thermofisher.com.
C6 is a glial cell line that was isolated from the brain of a rat with Glioma. The glial tumor was induced by N-nitrosomethylurea after a series of alternate culture and animal passages. This cell line was deposited by G. Sato and can be used in neuroscience and toxicology research.