Learn More
AdipoGen C6-L-HSL
SDP
Catalog No. 502560727
Encompass_Preferred
Chemical. CAS 147852-83-3. Formula C10H17NO3. MW 199.3. Synthetic. N-Hexanoyl-L-homoserine lactone is a small diffusible signaling molecule involved in quorum sensing, controlling gene expression and affecting cellular metabolism. The diverse applications of this molecule include regulation of virulence in general and in cystic fibrosis, infection prevention, slime and biofilm reduction in commercial agriculture and aquaculture industries, food spoilage prevention and septicemia in fish.
Catalog No. 50-256-0727 Supplier AdipoGen Supplier No. CDXH0111M050
Chemical. CAS 147852-83-3. Formula C10H17NO3. MW 199.3. Synthetic. N-Hexanoyl-L-homoserine lactone is a small diffusible signaling molecule involved in quorum sensing, controlling gene expression and affecting cellular metabolism. The diverse applications of this molecule include regulation of virulence in general and in cystic fibrosis, infection prevention, slime and biofilm reduction in commercial agriculture and aquaculture industries, food spoilage prevention and septicemia in fish.