Learn More
Cayman Chemical C6 L-threo Ceramide (d18:1/6:0), Purity: ≥98%, CAS: 189894-80-2, 1 mg
SDP
Catalog No. 502934661
Encompass_Preferred
A cell-permeable analog of naturally occurring ceramides; cytotoxic to U937 cells in vitro (IC50 = 18 μM); metabolically inactive; enhances PMA-induced IL-4 production in EL4 T cells when used at a concentration of 10 μM
Catalog No. 50-293-4661 Supplier Cayman Chemical Supplier No. 243901 MG
A cell-permeable analog of naturally occurring ceramides; cytotoxic to U937 cells in vitro (IC50 = 18 μM); metabolically inactive; enhances PMA-induced IL-4 production in EL4 T cells when used at a concentration of 10 μM