Learn More
Medchemexpress LLC C6 NBD ceramide | 94885-02-6 | MFCD00467024 | 99.3% | 575.74 g/mol | C30H49N5O6 | 1 MG
SDP
Catalog No. 5000285054
Encompass_Preferred
C6 NBD ceramide is a fluorescent short-chain ceramide analog used as a Golgi apparatus probe in live and fixed cells. It is cell-permeable and is metabolized to fluorescent sphingomyelin and glucosylceramide, enabling visualization of sphingolipid transport, Golgi morphology, and related cellular processes (Ex 466 nm, Em 536 nm).
  • Provides bright green fluorescence for imaging (Ex 466 nm, Em 536 nm).
  • Cell membrane permeable for live and fixed cell labeling.
  • Metabolized to fluorescent sphingomyelin and glucosylceramide for metabolism studies.
  • High purity (99.3%) for consistent experimental results.
  • Supplied as a 1 mg vial suitable for small-scale assays.
  • Store sealed at -20°C; in solvent, store at -80°C for up to 6 months or -20°C for up to 1 month.
Catalog No. 50-002-85054 Supplier Medchemexpress LLC Supplier No. HYW3561161MG
C6 NBD ceramide is a fluorescent short-chain ceramide analog used as a Golgi apparatus probe in live and fixed cells. It is cell-permeable and is metabolized to fluorescent sphingomyelin and glucosylceramide, enabling visualization of sphingolipid transport, Golgi morphology, and related cellular processes (Ex 466 nm, Em 536 nm).
  • Provides bright green fluorescence for imaging (Ex 466 nm, Em 536 nm).
  • Cell membrane permeable for live and fixed cell labeling.
  • Metabolized to fluorescent sphingomyelin and glucosylceramide for metabolism studies.
  • High purity (99.3%) for consistent experimental results.
  • Supplied as a 1 mg vial suitable for small-scale assays.
  • Store sealed at -20°C; in solvent, store at -80°C for up to 6 months or -20°C for up to 1 month.