Learn More
Abcam C6-NBD-ceramide, fluorescent sphingosine analog, 100UG
Catalog No. 502706034
Encompass_Preferred
MW 575.7 Da, Purity >98%. Fluorescent C6:0-sphingosine analog. For use in CERT trafficking studies and as internal standard. Possible activity in vitro. See similar compounds .
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6034 Supplier Abcam Supplier No. AB144090100UG
MW 575.7 Da, Purity >98%. Fluorescent C6:0-sphingosine analog. For use in CERT trafficking studies and as internal standard. Possible activity in vitro. See similar compounds .
The product is subject to the following: Abcam Restricted Use Statement