Learn More
Cayman Chemical C6 NBD CeramIded18 16 0 10mg
SDP
Catalog No. 502893580
Encompass_Preferred
C6 NBD ceramide is a biologically active fluorescent analog of short chain, membrane-permeable ceramides.{2754} It is as effective as C6 ceramide in the inhibition of viral glycoprotein transport through the Golgi.{2754} C6 NBD ceramide has been used as a fluorescent substrate for the activity of UDP-glucose:ceramide glucosyltransferase{2814} and to demonstrate the translocation of glucocerebroside and sphingomyelin from the Golgi to the plasma membrane.{3460}
Catalog No. 50-289-3580 Supplier Cayman Chemical Supplier No. 6252710 MG
C6 NBD ceramide is a biologically active fluorescent analog of short chain, membrane-permeable ceramides.{2754} It is as effective as C6 ceramide in the inhibition of viral glycoprotein transport through the Golgi.{2754} C6 NBD ceramide has been used as a fluorescent substrate for the activity of UDP-glucose:ceramide glucosyltransferase{2814} and to demonstrate the translocation of glucocerebroside and sphingomyelin from the Golgi to the plasma membrane.{3460}