Learn More
Cayman Chemical C6 NBD dihydro Ceramide (d18:0/6:0), Purity: ≥98%, CAS: 114301-97-2, 10 mg
SDP
Catalog No. 502890148
Encompass_Preferred
Structurally identical to C6 NBD ceramide, but contains a saturated bond in the C-4/C-5 position of the sphingosine backbone
Catalog No. 50-289-0148 Supplier Cayman Chemical Supplier No. 1000795710 MG
Structurally identical to C6 NBD ceramide, but contains a saturated bond in the C-4/C-5 position of the sphingosine backbone