Learn More
Cayman Chemical C6 NBD dihydro Ceramide (d18:0/6:0), Purity: ≥98%, CAS: 114301-97-2, 5 mg
SDP
Catalog No. 502887356
Encompass_Preferred
Structurally identical to C6 NBD ceramide, but contains a saturated bond in the C-4/C-5 position of the sphingosine backbone
Catalog No. 50-288-7356 Supplier Cayman Chemical Supplier No. 100079575 MG
Structurally identical to C6 NBD ceramide, but contains a saturated bond in the C-4/C-5 position of the sphingosine backbone