Learn More
Cayman Chemical C6 NBD Galactosylceramide d1
SDP
Catalog No. 502995487
Encompass_Preferred
A fluorescently tagged sphingolipid; tagged with C6 NBD; used to study the intracellular localization and metabolism of galatosylceramide
Catalog No. 50-299-5487 Supplier Cayman Chemical Supplier No. 22830100 UG
A fluorescently tagged sphingolipid; tagged with C6 NBD; used to study the intracellular localization and metabolism of galatosylceramide