Learn More
Medchemexpress LLC C6 NBD glucosylceramide | 94885-03-7 | MFCD00870345 | 98.0% | 737.88 g/mol | C36H59N5O11 | 500 UG
SDP
Catalog No. 5000285090
Encompass_Preferred
C6 NBD glucosylceramide is a fluorescently labeled short-chain glucosylceramide used as a probe to study sphingolipid metabolism, intracellular trafficking, and glucosylceramide synthase activity. It is provided as a yellow-orange solid for research use and exhibits strong NBD fluorescence for sensitive detection in biochemical and cell-based assays.
  • Fluorescent probe with excitation 466 nm and emission 535 nm.
  • High purity suitable for biochemical assays.
  • Short-chain analog for efficient cellular uptake.
  • Stable solid; store protected from moisture and light.
  • Compact 500 μg format for small-scale experiments.
Catalog No. 50-002-85090 Supplier Medchemexpress LLC Supplier No. HYW356117500UG
C6 NBD glucosylceramide is a fluorescently labeled short-chain glucosylceramide used as a probe to study sphingolipid metabolism, intracellular trafficking, and glucosylceramide synthase activity. It is provided as a yellow-orange solid for research use and exhibits strong NBD fluorescence for sensitive detection in biochemical and cell-based assays.
  • Fluorescent probe with excitation 466 nm and emission 535 nm.
  • High purity suitable for biochemical assays.
  • Short-chain analog for efficient cellular uptake.
  • Stable solid; store protected from moisture and light.
  • Compact 500 μg format for small-scale experiments.