Learn More
Cayman Chemical C6 NBD GlucosylcRamIDd18100ug
SDP
Catalog No. 502987604
Encompass_Preferred
A fluorescently tagged sphingolipid; tagged with C-6 NBD; used to study the metabolism and internalization of glucosylceramide
Catalog No. 50-298-7604 Supplier Cayman Chemical Supplier No. 23209100 UG
A fluorescently tagged sphingolipid; tagged with C-6 NBD; used to study the metabolism and internalization of glucosylceramide