Learn More
Cayman Chemical C6 NBD Lthreo CeramIded18100ug
SDP
Catalog No. 502933817
Encompass_Preferred
A fluorescently tagged sphingolipid; rapidly transferred between liposomes, labels the Golgi apparatus, and is metabolized to C6 NBD sphingomyelin in BHK cells and V79 fibroblasts
Catalog No. 50-293-3817 Supplier Cayman Chemical Supplier No. 24439100 UG
A fluorescently tagged sphingolipid; rapidly transferred between liposomes, labels the Golgi apparatus, and is metabolized to C6 NBD sphingomyelin in BHK cells and V79 fibroblasts