Learn More
Cayman Chemical C6 NBD PhytocRamIDt18 06 100ug
SDP
Catalog No. 502935958
Encompass_Preferred
A fluorescently tagged derivative of C6 phytosphingosine that is metabolized to fluorescent sphingomyelin and glucosylceramide in vitro
Catalog No. 50-293-5958 Supplier Cayman Chemical Supplier No. 24326100 UG
A fluorescently tagged derivative of C6 phytosphingosine that is metabolized to fluorescent sphingomyelin and glucosylceramide in vitro