Learn More
Carson C6 Series Lens Cleaner
SDPSCI_ED
Catalog No. S01586 Shop All Carson Optical Products
Change view
Click to view available options
Dimensions (L x W x H):
0.7 x 0.7 x 3.0 in.
0.9 x 0.7 x 5.1 in.
0.9 x 0.9 x 3.3 in.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W x H)
S01586 0.7 x 0.7 x 3.0 in.
S01587 0.9 x 0.9 x 3.3 in.
S01588 0.9 x 0.7 x 5.1 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. S01586 Supplier Carson Supplier No. CS35
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
Carson's C6 Lens Cleaner is designed to easily clean smudges, grime, and fingerprints from surfaces. It contains a special dry nano-particle cleaning formula that quickly removes these blemishes.
  • The C6 Lens Cleaner includes a retractable brush for dust and debris removal.
  • Its compact and sleek design enables easy transportation in a camera bag, purse, binocular pouch, or pocket.
  • The C6 Lens Cleaner is suitable for cleaning various types of lenses, including binoculars, spotting scopes, camcorders, and cameras.
  • It does not contain alcohol or ammonia, preventing damage to lens coatings.
  • A carbon-based reservoir is built into the lens cap.
  • To refill the lens cleaner tip with specially formulated carbon, push the cap back on and twist it.
Specifications
Type Lens Cleaner
Autoclavable Not Autoclavable
Description Carson C6 Series Reusable Compact Lens Cleaner with Dry Nano Particle Cleaning Formula (CS-35)
Dimensions (L x W x H) 0.7 x 0.7 x 3.0 in.
Size 13 mm (Tip)
Sterility Non-sterile
Voltage 0/1 V