Learn More
Cayman Chemical C6 Urea Ceramide, Purity: ≥95%, CAS: 486991-52-0, 10 mg
SDP
Catalog No. 503015669
Encompass_Preferred
An inhibitor of neutral ceramidase; It increases total ceramide levels in wild-type MEFs and HT-29 cells but not in MEFs lacking neutral ceramidase; inhibits proliferation of, and induces apoptosis and autophagy in HT-29, but not non-cancerous RIE-1, cells at 5 and 10 µM; reduces tumor growth and increases C16, C18, C20, and C24 ceramide levels in tumor tissue in an HT-29 mouse xenograft model at 1.25, 2.5, and 5 mg/kg for five days
Catalog No. 50-301-5669 Supplier Cayman Chemical Supplier No. 2549110 MG
An inhibitor of neutral ceramidase; It increases total ceramide levels in wild-type MEFs and HT-29 cells but not in MEFs lacking neutral ceramidase; inhibits proliferation of, and induces apoptosis and autophagy in HT-29, but not non-cancerous RIE-1, cells at 5 and 10 µM; reduces tumor growth and increases C16, C18, C20, and C24 ceramide levels in tumor tissue in an HT-29 mouse xenograft model at 1.25, 2.5, and 5 mg/kg for five days