Learn More
MSC Starrett 52682 36" Long, 1/64, 1/32, 1/16, 1/8" Graduation, Rigid Spring Steel Rule 4R Graduation Style, 1-1/4" Wide, Silver, Satin Chrome Finish
Catalog No. 501304789
Encompass
Starrett 52682 36" Long, 1/64, 1/32, 1/16, 1/8" Graduation, Rigid Spring Steel Rule 4R Graduation Style, 1-1/4" Wide, Silver, Satin Chrome Finish
Catalog No. 50-130-4789 Supplier MSC Supplier No. 86421369
Add to Cart
Add to Cart
Starrett 52682 36" Long, 1/64, 1/32, 1/16, 1/8" Graduation, Rigid Spring Steel Rule 4R Graduation Style, 1-1/4" Wide, Silver, Satin Chrome Finish