Learn More
MSC Starrett 52630 150mm Long, 0.5, 1mm Graduation, Rigid Spring Steel Rule Metric Graduation Style, 19mm Wide, Silver, Satin Chrome Finish
Catalog No. NC1741505
Encompass
Starrett 52630 150mm Long, 0.5, 1mm Graduation, Rigid Spring Steel Rule Metric Graduation Style, 19mm Wide, Silver, Satin Chrome Finish
Catalog No. NC1741505 Supplier MSC Supplier No. 58704412
Starrett 52630 150mm Long, 0.5, 1mm Graduation, Rigid Spring Steel Rule Metric Graduation Style, 19mm Wide, Silver, Satin Chrome Finish