Learn More
AdipoGen C646,Chemical. CAS: 328968-36-1
SDP
Supplier:  AdipoGen AGCR13508M001
Encompass_Preferred
C646, AdipoGen Life Sciences. Chemical. CAS: 328968-36-1. Formula: C24H19N3O6. MW: 445.4. Reversible cell permeable p300/CBP histone acetyltransferase (HAT) inhibitor. Specific inhibition to p300 (86%) compared to N-acetyltransferase, PCAF, GCN5, Rtt109, Sas or MOZ histone acetyltransferases (<10%). Cell growth inhibitor in melanoma and non-small-cell-lung (NSCL) human cancer cell lines.
Catalog No. 50-464-721