Learn More
AdipoGen C646
SDP
Catalog No. 502562339
Encompass_Preferred
Chemical. CAS 328968-36-1. Formula C24H19N3O6. MW 445.4. Reversible cell permeable p300/CBP histone acetyltransferase HAT inhibitor. Specific inhibition to p300 86% compared to N-acetyltransferase, PCAF, GCN5, Rtt109, Sas or MOZ histone acetyltransferases <10%. Cell growth inhibitor in melanoma and non-small-cell-lung NSCL human cancer cell lines.
Catalog No. 50-256-2339 Supplier AdipoGen Supplier No. AGCR13508M005
Chemical. CAS 328968-36-1. Formula C24H19N3O6. MW 445.4. Reversible cell permeable p300/CBP histone acetyltransferase HAT inhibitor. Specific inhibition to p300 86% compared to N-acetyltransferase, PCAF, GCN5, Rtt109, Sas or MOZ histone acetyltransferases <10%. Cell growth inhibitor in melanoma and non-small-cell-lung NSCL human cancer cell lines.