Learn More
Aviva Systems Biology Corp ZUP1 Antibody - middle region (ARP32274_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32274_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against C6orf113. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human C6orf113. Alternate names: RP3-412I7.3, dJ412I7.3, C6orf113ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-54507