Learn More
Aviva Systems Biology Corp C6orf134 Antibody - N-terminal region (ARP42641_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42641_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against C6orf134. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human C6orf134. Alternate names: DKFZp547J097, FLJ13158, Nbla00487, TAT, MEC17, C6orf134ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-62946