Learn More
Aviva Systems Biology Corp C6orf146 Antibody - middle region (ARP55647_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP55647_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against C6orf146. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human C6orf146. Alternate names: MGC43581, C6orf146ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-61564