Learn More
Aviva Systems Biology Corp CGAS Antibody - middle region (ARP52607_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP52607_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against C6orf150. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human C6orf150. Alternate names: MGC131892, MGC142166, MGC142168, C6orf150ApplicationWB. Species reactivity: Cow, Guinea Pig, Horse, Human, Pig, Rabbit
Catalog No. 50-014-60782