Learn More
Aviva Systems Biology Corp C6orf173 Antibody - N-terminal region (ARP54417_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP54417_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against C6orf173. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human C6orf173. Alternate names: CUG2, DKFZp686L1553, C6orf173ApplicationWB. Species reactivity: Guinea Pig, Horse, Human
Catalog No. 50-014-53738