Learn More
Aviva Systems Biology Corp C6orf182 Antibody - middle region (ARP55720_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP55720_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against C6orf182. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human C6orf182. Alternate names: MGC21731, MGC70837, bA487F23.2, cep57R, C6orf182ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast
Catalog No. 50-286-071