Learn More
Aviva Systems Biology Corp C6orf199 Antibody - middle region (ARP55457_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP55457_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against C6orf199. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human C6orf199. Alternate names: FLJ42177, MGC26954, RP1-70A9.1, dJ70A9.1, AKD2, C6orf199, C6orf224ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Rabbit, Rat
Catalog No. 50-014-61513