Learn More
Aviva Systems Biology Corp C6orf21 Antibody - middle region (ARP44450_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP44450_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against C6orf21. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human C6orf21. Alternate names: G6f, LY6G6D, NG32, C6orf21ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Rabbit
Catalog No. 50-014-58814