Learn More
Medchemexpress LLC C7/Complement compon 5ug
SDP
Catalog No. 5000178868
Encompass_Preferred
C7 also known as complement component C7 protein is a key component of the membrane attack complex (MAC) and plays a key role in innate and adaptive immune responses As a component of MAC C7 actively participates in the formation of target cell plasma membrane pores C7/Complement component C7 Protein Human (HEK293 Fc) is the recombinant human-derived C7/Complement component C7 protein expressed by HEK293 with C-hFc labeled tag
Catalog No. 50-001-78868 Supplier Medchemexpress LLC Supplier No. HYP743725UG
Add to Cart
Add to Cart
C7 also known as complement component C7 protein is a key component of the membrane attack complex (MAC) and plays a key role in innate and adaptive immune responses As a component of MAC C7 actively participates in the formation of target cell plasma membrane pores C7/Complement component C7 Protein Human (HEK293 Fc) is the recombinant human-derived C7/Complement component C7 protein expressed by HEK293 with C-hFc labeled tag