Learn More
AdipoGen C7-DL-HSL
SDP
Catalog No. 502560703
Encompass_Preferred
Chemical. CAS 106983-26-0. Formula C11H19NO3. MW 213.3. Synthetic. N-Heptanoyl-L-homoserine lactone C7-HSL is a small diffusible signaling molecule involved in quorum sensing, thereby controlling gene expression and affecting cellular metabolism. The diverse applications of this kind of molecule include regulation of virulence, infection prevention and septicemia in fish.
Catalog No. 50-256-0703 Supplier AdipoGen Supplier No. CDXH0201M050
Chemical. CAS 106983-26-0. Formula C11H19NO3. MW 213.3. Synthetic. N-Heptanoyl-L-homoserine lactone C7-HSL is a small diffusible signaling molecule involved in quorum sensing, thereby controlling gene expression and affecting cellular metabolism. The diverse applications of this kind of molecule include regulation of virulence, infection prevention and septicemia in fish.